Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Replication Termination Factor 2 Antibody (OTI1E8), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271934APCCY7
Description
Replication Termination Factor 2 Monoclonal specifically detects Replication Termination Factor 2 in Human samples. It is validated for Flow Cytometry.Specifications
Replication Termination Factor 2 | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
RTF2 | |
Full length recombinant protein of human C20orf43 (NP_057491) produced in HEK293T cells. | |
0.1 mL | |
51507.0 | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
OTI1E8 | |
Flow Cytometry | |
CDAO5, chromosome 20 open reading frame 43, HSPC164, hypothetical protein LOC51507, SHUJUN-3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction