Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reprimo Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984262
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4262 Supplier Bioss Supplier No. BS1885R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May be involved in the regulation of p53-dependent G2 arrest of the cell cycle. Seems to induce cell cycle arrest by inhibiting CDK1 activity and nuclear translocation of the CDC2 cyclin B1 complex.

Specifications

Antigen Reprimo
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene RPRM
Gene Accession No. Q9NS64
Gene Alias 2410012A13Rik; candidate mediator of the p53 dependent G2 arrest; candidate mediator of the p53-dependent G2 arrest; FLJ90327; Protein reprimo; REPRIMO; reprimo, TP53 dependant G2 arrest mediator candidate; reprimo, TP53 dependent G2 arrest mediator candidate; Rprm
Gene Symbols RPRM
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Reprimo.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56475
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.