Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
REPS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP23814725UL
Description
REPS1 Polyclonal specifically detects REPS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
REPS1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
Q96D71 | |
REPS1 | |
This antibody was developed against a recombinant protein corresponding to amino acids: LITHTSTSPQQIPEQPNFADFSQFEVFAASNVNDEQDDEAEKHPEVLPAEKASDPASSLRVAKTDSKTEEKTAASAPANVSKGTTPLAPPPKPVRRRLKSEDE | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
RALBP1, RALBP1 associated Eps domain containing 1, ralBP1-associated Eps domain-containing protein 1, RalBP1-interacting protein 1 | |
Rabbit | |
Affinity Purified | |
RUO | |
85021 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction