Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ REPS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551414
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551414 Supplier Invitrogen™ Supplier No. PA551414
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KCELPLPRFM MSKNDGEIRF GNPAELHGTK VQIPYLTTEK NSFKRMDDED KQQETQSPTM SPLASPPSSP PHYQRVPLSH GYSKLRSSAE QMHPAPYEAR QPLVQPEGSS SGGPGTKPLR HQASLIRSFS VERELQDNSS YPDEPWR Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

The product of this gene is part of a protein complex that regulates the endocytosis of growth factor receptors. The encoded protein directly interacts with a GTPase activating protein that functions downstream of the small G protein Ral. Its expression can negatively affect receptor internalization and inhibit growth factor signaling. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen REPS2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene REPS2
Gene Accession No. Q8NFH8
Gene Alias partner of Ral-binding protein 1; partner of RalBP1; POB1; RALBP1 associated Eps domain containing 2; RALBP1 associated Eps domain containing protein 2; ralBP1-associated Eps domain-containing protein 2; RalBP1-interacting protein 2; REPS2
Gene Symbols REPS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human REPS2. Recombinant protein control fragment (Product #RP-92028).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9185
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.