Learn More
Description
RepSox synthesized to Ancillary Material Grade also available.
For more information about how RepSox may be used, see our protocols: Highly Efficient Generation of CiPSCs from MEFs, Generation of β cells from hPSCs, Reprogramming MEFs into Skeletal Muscle Progenitors .
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 287.32 |
| Purity | >99% |
| Molecular Weight (g/mol) | 287.326 |
| Chemical Name or Material | RepSox |
Safety and Handling
| Recommended Storage | Store at +4°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

