Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RepSox
GREENER_CHOICESDP

Catalog No. 374250 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
37-425-0 50 mg
37-4210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 37-425-0 Supplier Tocris Bioscience™ Supplier No. 3742/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective inhibitor of TGF-βRI

RepSox is a potent and selective inhibitor of the TGF-β type I receptor/ALK5 (IC50 values are 4 and 23 nM for TGF-β type I receptor autophosphorylation and binding respectively). Selective for TGF-β type I receptor over a range of kinases, including p38 MAPK, JNK1 and GSK3 (IC50 > 16 μM). Enhances the efficiency of cellular reprogramming; replaces Sox2 by inducing Nanog expression.

RepSox synthesized to Ancillary Material Grade also available.

For more information about how RepSox may be used, see our protocols: Highly Efficient Generation of CiPSCs from MEFs, Generation of β cells from hPSCs, Reprogramming MEFs into Skeletal Muscle Progenitors .

Chemical Identifiers

CAS 446859-33-2
Molecular Formula C17H13N5

Specifications

Quantity 50 mg
Formula Weight 287.32
Purity >99%
Molecular Weight (g/mol) 287.326
Chemical Name or Material RepSox
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.