Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RER1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565659
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565659 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565659 Supplier Invitrogen™ Supplier No. PA565659
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSPKVDPSLME DSDDGPSLPT KQNEEFRPFI RRLPEFK Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

Involved in the retrieval of endoplasmic reticulum membrane proteins from the early Golgi compartment.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RER1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RER1
Gene Accession No. O15258
Gene Alias 1110060F11Rik; 5830454N22Rik; AU043380; Protein RER1; Rer1; RER1 homolog; rer1 protein; RER1 retention in endoplasmic reticulum 1 homolog; RER1 retention in endoplasmic reticulum 1 homolog (S. cerevisiae); retention in endoplasmic reticulum sorting receptor 1; RGD1306324; zgc:65968
Gene Symbols RER1
Host Species Rabbit
Immunogen Recombinant Human RER1. Recombinant protein control fragment (Product #RP-106335).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11079
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.