Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RERG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535964
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (98%), Xenopus laevis (89%), Rhesus Monkey (100%), Bovine (99%).
RERG, a member of the RAS superfamily of GTPases, inhibits cell proliferation and tumor formation.
Specifications
RERG | |
Polyclonal | |
Unconjugated | |
RERG | |
MGC15754; RAS like estrogen regulated growth inhibitor; RAS-like; RAS-like, estrogen-regulated, growth inhibitor; RAS-like, estrogen-regulated, growth-inhibitor; ras-related and estrogen-regulated growth inhibitor; ras-related and estrogen-regulated growth protein; RERG; RGD1562829; si:cabz01085950.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
502916, 85004 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q96A58 | |
RERG | |
Recombinant fragment corresponding to a region within amino acids 1 and 199 of RERG. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction