Learn More
Restek™ Res-Sil B Packings
SDP
Catalog No. 06712911 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Particle Size:
60/80 Mesh
80/100 Mesh
Material:
1% TCEP on ResSil B
ResSil B
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Material
06-712-911 60/80 Mesh ResSil B
06-712-650 80/100 Mesh ResSil B
06-712-651 80/100 Mesh 1% TCEP on ResSil B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-712-911 Supplier Restek™ Supplier No. 25401
Unique separation of saturated and unsaturated hydrocarbons.
  • Innovative bonding chemistry for batch-to-batch reproducibility, excellent thermal stability, and long life
  • Wide range of bonded phases available
  • Equivalent to Durapak and Porasil packings
Specifications
Particle Size 60/80 Mesh
Type Packing Material
Material ResSil B