Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Resazurin
SCI_ED

Catalog No. S25783 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 g
20 mL
Packaging:
Solid
Solution - 1%

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 62758-13-8
Molecular Formula C12H6NNaO4
Molecular Weight (g/mol) 251.173
InChI Key IVGPGQSSDLDOLH-UHFFFAOYSA-M
Synonym Biological Stain/Indicator, 7-Hydroxy-3H-phenoxazin-3-one 10-oxide, sodium salt
PubChem CID 112939
IUPAC Name sodium;10-oxido-7-oxophenoxazin-10-ium-3-olate
SMILES C1=CC2=C(C=C1[O-])OC3=CC(=O)C=CC3=[N+]2[O-].[Na+]

Specifications

Assay Percent Range >99% w/v
Linear Formula C12H6NO4Na
Formula Weight 251.17
Packaging Solution - 1%
Color Black
Quantity 20 mL
Chemical Name or Material Resazurin
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.