Learn More
Invitrogen™ Resazurin, Sodium Salt
Description
Resazurin, Sodium Salt
The non-fluorescent resazurin can be used as a fluorogenic oxidation-reduction indicator in a variety of cells, including bacteria, yeast and eukaryotes by flow cytometry, fluorescence microscopy and high-throughput screening. The red-fluorescent product, resorufin exhibits absorption/emission maxima ∼575/585 nm.
Specifications
Specifications
| Color | Red |
| Content And Storage | Store at room temperature and protect from light. |
| Description | Resazurin, Sodium Salt |
| Detection Method | Fluorescence |
| For Use With (Application) | Cell Viability |
| For Use With (Equipment) | Microplate Reader |
| Product Type | Resazurin |
| Dye Type | Resazurin |
| Form | Solid |
| Product Line | Invitrogen |
| Show More |
Frequently Asked Questions (FAQs)
alamarBlue reagent and PrestoBlue reagent contain resazurin in a proprietary stabilizing formulation that allows for a convenient mix, incubate, and read protocol. PrestoBlue reagent is an improvement in the formulation of alamarBlue reagent that allows for much faster staining (typically 10 minutes vs. 1-4 hours to obtain a similar signal and sensitivity). C12-resazurin is a derivative of resazurin that has better cellular retention and thus allows for analysis on a flow cytometer and multiplexing with viability indicators and other biomarkers.
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.