Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Research™ plus Mechanical Multi-Channel Pipette PROMO

Catalog No. 05413602PM2 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
1 to 20 μL
5 to 100 μL
Number of Channels:
16
24
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Number of Channels
05-413-602PM2 5 to 100 μL 24
05-413-599PM2 1 to 20 μL 16
05-413-601PM2 5 to 100 μL 16
05-413-600PM2 1 to 20 μL 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 05-413-602PM2 Supplier Eppendorf™ Supplier No. 2231001066

Save up to 30% on qualifying packages that include one Eppendorf™ Research™ plus Multichannel Mechanical Pipette and two packs of Eppendorf™ twin.tec™ 384-Well PCR Plates (Cat. No. E951020702).

Promo:

Save up to 30% on qualifying combination packages that include one Eppendorf™ Research™ plus Multichannel Mechanical Pipette and two packs (25 plates per pack) of Eppendorf™ twin.tec™ 384-Well PCR Plates (Cat. No. E951020702).

Choose a package that features a:

  • Research plus 16-Channel Pipette, 1 to 20 μL or 5 to 100 μL or
  • Research plus 24-Channel Pipette, 1 to 20 μL or 5 to 100 μL

NOTE: Savings based on the cost of purchasing the products separately. Discounts taken off list price.

Research™ plus Multichannel Mechanical Pipette Features

  • Volume adjustment feature allows desired volume to be reached in few quick turns
  • Control button and ejector's low operating force are positioned for ergonomics
  • 4-digit, magnifying-shape volume disp
  • Calibration display and adjustment opening for specific liquids and volumes
  • Has ability to be autoclaved without disassembling

twin.tec™ 384-Well PCR Plates (Cat. No. E951020702) Features

  • One-piece design: combining a polycarbonate frame and polypropylene wells for optimum performance.
  • Extremely thin-walled polypropylene wells guarantee optimum heat transfer to the sample.
  • Exceptionally solid and torque-resistant polycarbonate frame.
  • Cut-away corner and alphanumeric grid-referencing.
  • Raised well rims for effective sealing, also reduces risk of cross-contamination.
  • Batch-tested and certified free of DNA, DNase, RNase and PCR inhibitors (PCR clean)
TRUSTED_SUSTAINABILITY

Specifications

Battery Type None
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Light Yellow
Number of Channels 24
Product Line Research Plus
Type Mechanical Pipette
Volume (Metric) 5 to 100 μL
Display 4 Digits
Finger Style Ergonomic
Includes Box 2.0 with 384 pipette tips and 2 free boxes 384-well, clear, Eppendorf twin.tec™ skirted PCR plates (25 plates per box)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.