Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Reserpine
GREENER_CHOICESDP

Catalog No. 27421G Shop All Novus Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-421-G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 27-421-G Supplier R&D Systems™ Supplier No. 2742/1G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Inhibitor of vesicular monoamine transport

Binds the vesicular monoamine transporter (VMAT2) and inhibits transport of biogenic amines into adrenal chromaffin granules and synaptic vesicles. Causes depletion of biogenic amine stores. Antihypertensive and antipsychotic.

Chemical Identifiers

CAS 50-55-5
Molecular Formula C33H40N2O9

Specifications

Quantity 1 g
Product Line Tocris
Formula Weight 608.68
Purity >99%
Molecular Weight (g/mol) 608.688
Chemical Name or Material Reserpine
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.