Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Resiquimod
GREENER_CHOICE

Catalog No. 453610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-361-0 10 mg
45-365-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 45-361-0 Supplier Tocris Bioscience™ Supplier No. 4536/10
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Toll-like receptor 7 (TLR7) agonist

Toll-like receptor 7 (TLR7) agonist. Induces upregulation of IL-6, IL-12, IFN-γ and iNOS expression in mouse bone marrow-derived macrophages (BMMs). Inhibits RANKL-induced osteoclast differentiation in mouse BMMs and human peripheral blood monocytes. Also promotes the differentiation of MDSCs into macrophages and dendritic cells. Proinflammatory and antiviral.

Chemical Identifiers

CAS 144875-48-9
Molecular Formula C17H22N4O2

Specifications

Quantity 10 mg
Formula Weight 314.38
Purity >98%
Molecular Weight (g/mol) 314.389
Chemical Name or Material Resiquimod
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.