Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Resistin Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
| Antigen | Resistin |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Resistin Polyclonal specifically detects Resistin in Human samples. It is validated for Western Blot.Specifications
| Resistin | |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Rabbit | |
| Diabetes Research | |
| PBS with 0.02% Sodium Azide | |
| 56729 | |
| Reacts with residues 94-102 [AGMDWTGAR] of the human Resistin protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Adipose tissue-specific secretory factor, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN1, RSTNXCP1 | |
| RETN | |
| IgG | |
| Affinity Purified | |
| Reacts with residues 94-102 [AGMDWTGAR] of the human Resistin protein. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title