Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Resistin Antibody (MRES18), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP280087B
This item is not returnable.
View return policy
Description
Resistin Monoclonal antibody specifically detects Resistin in Mouse samples. It is validated for ELISASpecifications
Resistin | |
Monoclonal | |
Biotin | |
Adipose tissue-specific secretory factor, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN1, RSTNXCP1 | |
Recombinant mouse resistin. | |
0.1 mL | |
Diabetes Research | |
56729 | |
Store at 4°C in the dark. | |
IgG1 κ |
ELISA | |
MRES18 | |
PBS | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction