Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Resistin Mouse anti-Human, Clone: RETN/4324, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | Resistin |
---|---|
Clone | RETN/4324 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Description
Resistin Monoclonal antibody specifically detects Resistin in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)Specifications
Resistin | |
0.2 mg/ml | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Diabetes Research | |
10 mM PBS with 0.05% BSA | |
Adipose tissue-specific secretory factor, ADSFMGC126609, C/EBP-epsilon regulated myeloid-specific secreted cysteine-rich proteinprecursor 1, C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein, Cysteine-rich secreted protein A12-alpha-like 2, Cysteine-rich secreted protein FIZZ3, FIZZ3MGC126603, found in inflammatory zone 3, HXCP1, resistin, RETN1, RSTNXCP1 | |
Recombinant fragment of human Resistin protein (around aa 19-108) (exact sequence is proprietary) (Uniprot: Q9HD89) | |
Primary | |
Store at 4°C. |
RETN/4324 | |
Western Blot 2-4 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q9HD89 | |
56729 | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title