Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resistin Rabbit anti-Rat, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. AB3371I25UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3371-I-25UL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3371-I-25UL Supplier MilliporeSigma Supplier No. AB3371I25UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Resistin Antibody, Cat. No. AB3371-I, is a rabbit polyclonal antibody that detects rat resistin and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Resistin
Applications Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Adipose tissue-specific secretory factor;ADSF;C/EBP-epsilon-regulated myeloid-specific secreted cysteine-rich protein;Cysteine-rich secreted protein A12-alpha-like 2;Cysteine-rich secreted protein FIZZ3
Gene Symbols RETN;FIZZ3;HXCP1;RSTN
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal region of rat resistin.
Molecular Weight of Antigen ∽12 kDa observed; 11.42 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Inflammation & Immunology
Primary or Secondary Primary
Test Specificity This rabbit polyclonal antibody detects Resistin in rat tissues. It targets an epitope within 17 amino acids from the N-terminal region.
Target Species Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.