Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Resorcinol
SCI_ED

Catalog No. p-4443146 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Powder

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O

Specifications

Color White
Density 1.272g/mL
Assay Percent Range >99% w/v
Packaging Powder
Linear Formula C6H4-1, 3-(OH)2
Quantity 25 g
Formula Weight 110.11
Chemical Name or Material Resorcinol
ShelfLife 5 years
DOTInformation DOT Class 6.1, III, : Poison
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.