Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resorcinol, MP Biomedicals™

Catalog No. ICN10280891 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10280891 1 kg
02-102-80890 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10280891 Supplier MP Biomedicals, Inc Supplier No. 0210280891

Resorcinol is an aromatic alcohol used in large scale processes, dermatology research, and in colorimetric assays for sialic acid.

  • Resorcinol is an aromatic alcohol used in large scale processes such as tanning, the manufacture of resins and resin adhesives, the production of hexylresorcinol and p-aminosalicylic acid, and the dyeing and printing of textiles.
  • Resorcinol is used in dermatology research, such as in studies of irritant contact dermatitis.
  • The role of resorcinol in causing cytotoxicity and cytokine release in human epidermal keratinocytes and dermal fibroblasts has been studied.
  • The cytoxicity of resorcinol and other phenolic compounds against the melanotic human melanoma cell lines IRE 1 and IRE 2, and the lymphoma- and leukemia-derived cell lines Raji and K 562 has been studied, at phenolic compound concentrations of 10 μM to 5 mM.
  • The use of resorcinol in a colorimetric assay for sialic acid has been reported.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00002269
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O

Specifications

Melting Point 228 to 232°F
Color White
Density 1.2 g/cm3 at 68°F
Boiling Point 531°F to 536°F (at 760 mm Hg)
Flash Point 261°F
Ignition Point 1125°F
Assay Percent Range ≥99%
Quantity 1 kg
Solubility Information ≥100 mg/mL at 65.3°F
Vapor Density 3.79
Vapor Pressure 1 mm Hg at 227.1°F, 0.0002 mm Hg at 77°F
Formula Weight 110.11
Percent Purity ≥99%
Physical Form Solid
For Use With (Application) Building Blocks, Chemical Synthesis
Chemical Name or Material Resorcinol, Molecular Biology, Cell Biology
Show More Show Less
EINECSNumber 203-585-2
Recommended Storage Store at room temperature.

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.