Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resorcinol, ACS, 99.0-100.5%

Catalog No. aa3624822
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 108-46-3 | C6H6O2 | 110.112 g/mol

Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-46-3
Molecular Formula C6H6O2
Molecular Weight (g/mol) 110.112
MDL Number MFCD00002269
InChI Key GHMLBKRAJCXXBS-UHFFFAOYSA-N
Synonym resorcinol, 1,3-benzenediol, resorcin, 1,3-dihydroxybenzene, 3-hydroxyphenol, m-hydroquinone, resorcine, m-dihydroxybenzene, m-hydroxyphenol, m-benzenediol
PubChem CID 5054
ChEBI CHEBI:27810
IUPAC Name benzene-1,3-diol
SMILES C1=CC(=CC(=C1)O)O

Specifications

Melting Point 109°C to 112°C
Density 1.272 g/mL
Boiling Point 280°C to 281°C
Flash Point 171°C (339°F)
Assay Percent Range 99.0 to 100.5%
Quantity 100 g
UN Number UN2876
Beilstein 906905
Sensitivity Air and light sensitive
Merck Index 14,8155
Solubility Information Soluble in water,alcohol,ether and glycerol.
Formula Weight 110.11
Percent Purity 99.0 to 100.5%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Resorcinol, For analysis ACS
Show More Show Less
Hazard Category H301-H315-H319-H500
Hazard Statement GHS H Statement
H302-H315-H319
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: RESORCINOL
EINECSNumber 203-585-2
RTECSNumber VG9625000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.