Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Resorcinol, Flake, Technical, Spectrum™ Chemical
SDP

Catalog No. 18613024 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
25 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-024 2.5 kg Poly Pail
18-613-026 500 g Poly Pail
18-613-025 25 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-613-024 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. R15292.5KG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-46-3

Specifications

CAS Max % 1
Boiling Point 277 to 281°C
Packaging Poly Pail
Quantity 2.5 kg
Grade Technical
Physical Form Crystalline Powder or Lumps

CAUTION: For manufacturing or laboratory use only. Not for drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.