Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RESP18 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561108
Description
Immunogen sequence: PTKTTESPLA KVNRDQCFTS EVVSKALKQE VANPVKITYR CSYGGLDMMQ APGPSKEEII YKIMRLLWAT S Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 28%.

Specifications
RESP18 | |
Polyclonal | |
Unconjugated | |
RESP18 | |
regulated endocrine specific protein 18; regulated endocrine-specific protein 18; regulated endocrine-specific protein 18 homolog; RESP18 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
389075 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5W5W9 | |
RESP18 | |
Recombinant protein corresponding to Human RESP18. Recombinant protein control fragment (Product #RP-96212). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction