Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus fusion protein Antibody (10B6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063020
Description
Respiratory Syncytial Virus fusion protein Monoclonal specifically detects Respiratory Syncytial Virus fusion protein in Virus samples. It is validated for Flow Cytometry, ELISA.Specifications
Respiratory Syncytial Virus fusion protein | |
Monoclonal | |
1.0 mg/mL | |
Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
Mouse | |
Protein A or G purified | |
RUO | |
Recognises the Respiratory Syncytial Virus (RSV) Fusion protein, also known as F-protein. The F-protein is a membrane-associated protein found in the membrane of infected respiratory epithelial cells. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b |
Flow Cytometry, ELISA | |
10B6 | |
Unconjugated | |
RSV fusion protein | |
Purified Respiratory Syncytial Virus (RSV) | |
0.1 mg | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction