Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus fusion protein Antibody (RSV3216 (B016)), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064495AF350
Description
Respiratory Syncytial Virus fusion protein Monoclonal antibody specifically detects Respiratory Syncytial Virus fusion protein in Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
Respiratory Syncytial Virus fusion protein | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
RSV3216 (B016) | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
RSV fusion protein | |
Bovine Respiratory Syncytial Virus fusion protein strains: 127, SNK and 9007. Human RSV strains: Long, Randall, 8/60, and A/2. | |
0.5 mL | |
Primary | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction