Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus Glycoprotein F Antibody (11-2-C9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2501560.025ML
Description
Respiratory Syncytial Virus Glycoprotein F Monoclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Functional AssaySpecifications
Respiratory Syncytial Virus Glycoprotein F | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Functional | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Western Blot, ELISA, Immunofluorescence, Functional Assay | |
11-2-C9 | |
Unconjugated | |
PBS | |
Gradient-purified RSF-44 virus (subgroup A) UV inactivated for 20 minutes at 20C | |
0.025 mL | |
Virology Bacteria and Parasites | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction