Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus Glycoprotein F Antibody (11-3-A3), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250392UV
Description
Respiratory Syncytial Virus Glycoprotein F Monoclonal specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Respiratory Syncytial Virus Glycoprotein F | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Gradient-purified RSF-44 virus (subgroup A) UV inactivated for 20 minutes at 20C | |
0.1 mL | |
Virology Bacteria and Parasites | |
The antibody reacts with the RS virus F1 protein from both subgroups A and B of the virus. | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
11-3-A3 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction