Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus Glycoprotein F Antibody (11-6-F9), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250394AF405
Description
Respiratory Syncytial Virus Glycoprotein F Monoclonal specifically detects Respiratory Syncytial Virus Glycoprotein F in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Respiratory Syncytial Virus Glycoprotein F | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Gradient-purified RSF-44 virus (subgroup A) UV inactivated for 20 minutes at 20C | |
0.1 mL | |
Virology Bacteria and Parasites | |
Immunoblot using the reduced and unreduced RS virus shows that 11-6-F9 reacts with the non-reduced form of the virus F protein (VP70). It has a plaque reduction neutralization titer of 10 1.3. Induce in vitro antibody-dependent enhancement of macrophage cells.This antibody has a RS virus 90% plaque neutralization titre of 101.3.and it also has RSV enhancing activity (enhancement index range: 2.5-5.3). Gimenez et al. (1996). Gimenez et al. (1996) was the first report showing that a monoclonal antibody specific for the F protein has enhancing activity. | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
11-6-F9 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction