Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus Glycoprotein G Antibody (9H2G5C6), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306593AF700
Description
Respiratory Syncytial Virus Glycoprotein G Monoclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein G in Virus samples. It is validated for Western Blot, ELISASpecifications
Respiratory Syncytial Virus Glycoprotein G | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant HumanRespiratory Syncytial Virus Glycoprotein G (His 67-Arg 297; P27022-1). | |
0.1 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
9H2G5C6 | |
Western Blot, ELISA | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction