Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Respiratory Syncytial Virus Glycoprotein G Antibody, PE, Novus Biologicals™
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318801PE
Description
Respiratory Syncytial Virus Glycoprotein G Polyclonal antibody specifically detects Respiratory Syncytial Virus Glycoprotein G in Virus samples. It is validated for Western Blot, ELISASpecifications
| Respiratory Syncytial Virus Glycoprotein G | |
| Polyclonal | |
| Western Blot, ELISA | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified | 
| Western Blot, ELISA | |
| PE | |
| PBS | |
| Produced in rabbits immunized with purified, recombinant Respiratory Syncytial Virus Glycoprotein G (Accession#: AII22117.1; Ser64-Lys321). | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction