Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus Nucleoprotein Antibody (1C3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Respiratory Syncytial Virus Nucleoprotein |
---|---|
Clone | 1C3 |
Concentration | 1 mg/mL |
Dilution | ELISA, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
Applications | ELISA, Immunohistochemistry, Immunofluorescence |
Description
Respiratory Syncytial Virus Nucleoprotein Monoclonal antibody specifically detects Respiratory Syncytial Virus Nucleoprotein in Virus samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
Respiratory Syncytial Virus Nucleoprotein | |
1 mg/mL | |
ELISA, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Virology Bacteria and Parasites | |
PBS | |
IgG2a | |
Protein A purified |
1C3 | |
ELISA, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Virus | |
hRSV strain A2 infected HeLa cells | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title