Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus Nucleoprotein Antibody (1C3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250534H
Description
Respiratory Syncytial Virus Nucleoprotein Monoclonal specifically detects Respiratory Syncytial Virus Nucleoprotein in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
Respiratory Syncytial Virus Nucleoprotein | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
hRSV strain A2 infected HeLa cells | |
0.1 mL | |
Virology Bacteria and Parasites | |
This antibody has broad specificity for hRSV of both sub-groups A and B. | |
Store at 4C in the dark. | |
IgG2a |
ELISA, Immunohistochemistry | |
1C3 | |
ELISA, Immunohistochemistry | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction