Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus Nucleoprotein Antibody (RSV3132 (B023)), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10064752DL594
Description
Respiratory Syncytial Virus Nucleoprotein Monoclonal antibody specifically detects Respiratory Syncytial Virus Nucleoprotein in Human, Virus samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)Specifications
Respiratory Syncytial Virus Nucleoprotein | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Bovine RSV strains: 127, SNK and 9007. Human RSV strains: A2, Long, Randall and 8/60. | |
0.5 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
RSV3132 (B023) | |
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
Mouse | |
Protein A or G purified | |
RUO | |
Human, Virus | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction