Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus Subgroup A G Glycoprotein Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP318799IR
Description
Respiratory Syncytial Virus Subgroup A G Glycoprotein Polyclonal antibody specifically detects Respiratory Syncytial Virus Subgroup A G Glycoprotein in Virus samples. It is validated for Western Blot, ELISASpecifications
Respiratory Syncytial Virus Subgroup A G Glycoprotein | |
Polyclonal | |
Western Blot, ELISA | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Virus | |
Purified |
Western Blot, ELISA | |
DyLight 755 | |
50mM Sodium Borate | |
Produced in rabbits immunized with purified recombinant Human Respiratory Syncytial Virus Subgroup A G Glycoprotein (His67-Gln298, Uniprot: P20895-1) | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction