Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Respiratory Syncytial Virus Subgroup A G Glycoprotein Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB274755 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274755 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274755 Supplier Novus Biologicals Supplier No. NBP318799JF646
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Respiratory Syncytial Virus Subgroup A G Glycoprotein Polyclonal antibody specifically detects Respiratory Syncytial Virus Subgroup A G Glycoprotein in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Respiratory Syncytial Virus Subgroup A G Glycoprotein
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified recombinant Human Respiratory Syncytial Virus Subgroup A G Glycoprotein (His67-Gln298, Uniprot: P20895-1)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.