Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Respiratory Syncytial Virus Subgroup B Antibody (4-14), Novus Biologicals™

Mouse Monoclonal Antibody
$173.10
Specifications
Antigen | Respiratory Syncytial Virus Subgroup B |
---|---|
Clone | 2024-04-14 00:00:00 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunofluorescence |
Description
Respiratory Syncytial Virus Subgroup B Monoclonal antibody specifically detects Respiratory Syncytial Virus Subgroup B in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
Respiratory Syncytial Virus Subgroup B | |
1 mg/mL | |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Mouse | |
Virology Bacteria and Parasites | |
PBS | |
IgM | |
Thiophilic Chromatography |
2024-04-14 00:00:00 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Virus | |
The immunogen was gradient-purified RSN-2 virus (subgroup B) that was then treated with 0.1% SDS at 100C for 2 min. The procedure used to produce this antibody is described in Gimenez et al. (1984). | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title