Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

REST Mouse anti-Human, MilliporeSigma™

Catalog No. MABC156025 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1560-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1560-25 Supplier MilliporeSigma Supplier No. MABC156025UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect REST using this mouse monoclonal Anti-REST, clone 2D9.1 Antibody validated for use in Western Blotting and Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen REST
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols REST,NRSF,XBR
Host Species Mouse
Immunogen GST/His-Tagged recombinant fragment corresponding to 297 amino acids from the C-terminal region of human RE1-silencing transcription factor (REST).
Purification Method Protein G Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 2D9.1 detects RE1-silencing transcription factor in human cells. It targets an epitope within 297 amino acids from the C-terminal region.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.