Learn More
Restek™ ResTape PTFE Thread Tape
SDP
Catalog No. 06611087 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
For Use With (Application):
Gas Service
Oxygen Service
Stainless Steel
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
06-611-087 Gas Service
06-611-086 Oxygen Service
06-611-088 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 06-611-087 Supplier Restek™ Supplier No. 22486
Add to Cart
Add to Cart
For threaded connections in a wide range of plumbing materials
  • Available for gas, oxygen, and stainless steel services
  • Roll Dimensions: 260 x 1/2 in.
  • Maximum temperature: 260°C
Specifications
Type Tape
Material PTFE
Description Color: Yellow
For Use With (Application) Gas Service
Operating Temperature (Metric) Max: 260°C
Quantity 1 Roll