Learn More
Restek™ 1000 Series Manual Gas Tight Syringes (Hamilton)
SDP
Catalog No. 06712371 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Volume (Metric):
2.5 mL
Catalog No. Volume (Metric)
06-712-371 2.5 mL
1 options
Catalog No. 06-712-371 Supplier Restek™ Supplier No. 24581
Suitable for gases or liquids, for maximum inertness.
  • High accuracy of dispensed volumes
  • Interchangeable barrels, plungers, and tips extend performance and increase cost-effectiveness
Specifications
Material PTFE (Tip)
Needle Gauge 22 G
For Use With (Application) For gases or liquids
Length (English) Needle 2 in.
Product Type Hamilton PTFE Tip, Gas-Tight Syringes
Model 1002
Needle Point Number 2
Volume (Metric) 2.5 mL
Needle Point Style 2
Connection Type Cemented