Learn More
Restek™ 7microm In-Line Filter
SDP
Catalog No. p-3180043 Shop All Restek Corporation Products
Change view
Click to view available options
Material:
Siltek
Stainless Steel
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
06-712-265 Siltek
06-712-266 Stainless Steel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-712-265 Supplier Restek™ Supplier No. 24265
Only null left
Add to Cart
Add to Cart
Designed to collect particles larger than 7 microns.
  • Available in Siltek and stainless steel versions
  • With 0.25 in. compression fitting on both ends
Specifications
Type Filter
Porosity 7 μm
Material Siltek