Learn More
Restek™ Ignitor for Agilent GCs
SDP
Catalog No. 06718339 Shop All Restek Corporation Products
Encompass
Change view
Click to view available options
Packaging Quantity:
1 Each
Catalog No. Packaging Quantity
06-718-339 1 Each
1 options
Catalog No. 06-718-339 Supplier Restek™ Supplier No. 21001
Add to Cart
Edge
Add to Cart
For use with Agilent 5890, 6850, 6890, 7890 GCs.
  • Also fits OI Analytical 4410 detector
  • Quantity: 1 Each
Specifications
Type FID Ignitor
Packaging Quantity 1 Each
For Use With Agilent 5890, 6850, 6890, 7890 GCs