Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ ASI Check Valves for Waters Alliance Model 2690
SDP

For Waters Alliance Model 2690 and 2695.

Specifications

For Use With (Equipment) Waters Alliance Model 2690 and 2695
Type Inlet and Outlet Cartridge
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
06-712-716
SDP
View Documents
Restek™
25160
Sapphire and Ruby Ball and Seat N/A N/A N/A
06-712-718
SDP
View Documents
Restek™
25162
Ceramic Ball and Seat N/A N/A N/A
Description

Description

  • Valves do not leak even at pressures up to 12,000psi (82,737 kPa)
  • Valves close properly even without a high backpressure force
  • Cartridge design allows easy, convenient replacement of worn or damaged valves
Specifications

Specifications

Waters Alliance Model 2690 and 2695
Inlet and Outlet Cartridge
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.