Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Bonded Packed Column Stationary Phases: 10% Rtx-1
SDP

Catalog No. 06714445 Shop All Restek Corporation Products
Change view
Click to view available options
Type:
Agilent Configuration
General Configuration
Gow Mac 580
PE Auto Sys Configuration
PE/Sigma Configuration
SRI 8610C
Varian Configuration
Packing Material:
10% Rtx™-1
Silcoport™ W
Material:
SilcoSmooth™
Siltek-treated Stainless Steel
Stainless Steel
Stationary Phase:
10% Rtx™-1
Bonded Phase 10% Rtx™-1
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packing Material Material Stationary Phase
06-714-445 General Configuration Silcoport™ W SilcoSmooth™ 10% Rtx™-1
06-714-446 Agilent Configuration Silcoport™ W SilcoSmooth™ 10% Rtx™-1
06-714-589 General Configuration Silcoport™ W Stainless Steel 10% Rtx™-1
06-714-590 Agilent Configuration Silcoport™ W Stainless Steel 10% Rtx™-1
06-714-591 PE Auto Sys Configuration Silcoport™ W Stainless Steel 10% Rtx™-1
06-714-447 Varian Configuration Silcoport™ W SilcoSmooth™ 10% Rtx™-1
06-714-448 PE/Sigma Configuration Silcoport™ W SilcoSmooth™ 10% Rtx™-1
06-714-449 PE Auto Sys Configuration Silcoport™ W SilcoSmooth™ 10% Rtx™-1
06-714-450 Gow Mac 580 10% Rtx™-1 Siltek-treated Stainless Steel Bonded Phase 10% Rtx™-1
06-714-451 SRI 8610C 10% Rtx™-1 Siltek-treated Stainless Steel Bonded Phase 10% Rtx™-1
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. 06-714-445 Supplier Restek™ Supplier No. 80405800

Bonded phases in capillary columns provide a dramatic increase in column quality. Restek Bonded Packed Column Stationary Phases: 10% Rtx-1 are for on-column injections.

  • Low bleed levels
  • Longer column lifetimes
  • Short conditioning times due to solvent rinsing during manufacturing process which removes low molecular weight polymer fragments
  • Bonded packings usually stabilize in 30 minutes or less
  • Detectors can be operated at higher sensitivities due to low-bleed
  • Reduces need for data system bleed compensation
  • More resistant to oxidation than nonbonded phases because of the stronger intermolecular forces produced by cross-linking
  • No sagging due to thorough cross-linking of material
  • Mesh: 100/120 Silcoport W

Order Info

Initial 2 in. of column will be empty, to accommodate a needle. For a completely filled column add suffix -901. Agilent configuration also includes 1.5 in. void on detector side.

Column Configuration Key: General - General Configuration; Agilent - Agilent 5880, 5890, 5987, 6890, 7890; PE Auto Sys - PE Auto System 8300, 8400, 8700, Clarus 500.

Specifications

Film Thickness 2 μm
Particle Size 2 μm
Stationary Phase 10% Rtx™-1
USP Type G1
Type General Configuration
Max. Temperature 380/400°C
For Use With (Application) High Bioling Compounds, Crude Oils, Triglycerides, Waxes
Temperature Range 0°C to 350°C
Length (Metric) 2 m
Material SilcoSmooth™
Packing Material Silcoport™ W
Mesh Size 100/120
Diameter (Metric) Inner 2 mm
Diameter (Metric) Outer 3.17 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.