Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Bonded Packed Column Stationary Phases: 20% Rtx-Stabilwax
SDP

Catalog No. 06714605 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Product Type:
Agilent Configuration
General Configuration
PE Auto Sys Configuration
PE/Sigma Configuration
Shimadzu 8A
Varian Configuration
Packing Material:
20% Rtx™-Stabilwax
Silcoport™ W
Material:
SilcoSmooth™
Stainless Steel
Stationary Phase:
20% Rtx™-Stabilwax
Bonded Phase 20% Rtx™-Stabilwax (Carbowax™ Phase)
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Packing Material Material Stationary Phase
06-714-605 Shimadzu 8A 20% Rtx™-Stabilwax Stainless Steel Bonded Phase 20% Rtx™-Stabilwax (Carbowax™ Phase)
06-714-604 Agilent Configuration Silcoport™ W Stainless Steel 20% Rtx™-Stabilwax
06-714-500 Agilent Configuration Silcoport™ W SilcoSmooth™ 20% Rtx™-Stabilwax
06-714-501 Varian Configuration Silcoport™ W SilcoSmooth™ 20% Rtx™-Stabilwax
06-714-499 General Configuration Silcoport™ W SilcoSmooth™ 20% Rtx™-Stabilwax
06-714-503 PE Auto Sys Configuration Silcoport™ W SilcoSmooth™ 20% Rtx™-Stabilwax
06-714-502 PE/Sigma Configuration Silcoport™ W SilcoSmooth™ 20% Rtx™-Stabilwax
06-714-603 General Configuration Silcoport™ W Stainless Steel 20% Rtx™-Stabilwax
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 06-714-605 Supplier Restek™ Supplier No. 80449851
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bonded phases in capillary columns provide a dramatic increase in column quality. Restek Bonded Packed Column Stationary Phases: 20% Rtx-Stabilwax are for on-column injections.

  • Low bleed levels
  • Longer column lifetimes
  • Short conditioning times due to solvent rinsing during manufacturing process which removes low molecular weight polymer fragments
  • Bonded packings usually stabilize in 30 minutes or less
  • Detectors can be operated at higher sensitivities due to low-bleed
  • Reduces need for data system bleed compensation
  • More resistant to oxidation than nonbonded phases because of the stronger intermolecular forces produced by cross-linking
  • No sagging due to thorough cross-linking of material
  • Mesh: 100/120 Silcoport W

Order Info

Initial 2 in. of column will be empty, to accommodate a needle. For a completely filled column add suffix -901. Agilent configuration also includes 1.5 in. void on detector side.

Column Configuration Key: General - General Configuration

Specifications

Max. Temperature 240°C
For Use With (Application) Xylene Isomers Analysis, Flavor Compounds, Essential Oils, Solvents, Xylene Isomers, US EPA Method 603
Temperature Range 40°C to 260°C
Length (Metric) 1828.8 mm
Material Stainless Steel
Packing Material 20% Rtx™-Stabilwax
Mesh Size 100/120
Diameter (Metric) Inner 2 mm
Diameter (Metric) Outer 3.17 mm
USP Type G14, G15, G16, G20, G39
Stationary Phase Bonded Phase 20% Rtx™-Stabilwax (Carbowax™ Phase)
Product Type Shimadzu 8A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.