Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Bonded Packed Column Stationary Phases: 3% Rtx-5
SDP

Catalog No. 06714596 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
General Configuration
Shimadzu 8A
Varian 3700, Vista Series, FID
Packing Material:
3% Rtx™-5
Silcoport™ W
Material:
SilcoSmooth™
Siltek-treated Stainless Steel
Stainless Steel
Stationary Phase:
3% Rtx™-5
Bonded Phase 3% Rtx™-5
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packing Material Material Stationary Phase
06-714-596 Varian 3700, Vista Series, FID 3% Rtx™-5 Stainless Steel Bonded Phase 3% Rtx™-5
06-714-672 General Configuration Silcoport™ W SilcoSmooth™ 3% Rtx™-5
06-714-673 Varian 3700, Vista Series, FID 3% Rtx™-5 Siltek-treated Stainless Steel Bonded Phase 3% Rtx™-5
06-714-674 Shimadzu 8A 3% Rtx™-5 Siltek-treated Stainless Steel Bonded Phase 3% Rtx™-5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-714-596 Supplier Restek™ Supplier No. 80444820901

Bonded phases in capillary columns provide a dramatic increase in column quality. Restek Bonded Packed Column Stationary Phases: 3% Rtx-5 are for on-column injections.

  • Low bleed levels
  • Longer column lifetimes
  • Short conditioning times due to solvent rinsing during manufacturing process which removes low molecular weight polymer fragments
  • Bonded packings usually stabilize in 30 minutes or less
  • Detectors can be operated at higher sensitivities due to low-bleed
  • Reduces need for data system bleed compensation
  • More resistant to oxidation than nonbonded phases because of the stronger intermolecular forces produced by cross-linking
  • No sagging due to thorough cross-linking of material
  • Mesh: 100/120 Silcoport W

Order Info

Initial 2 in. of column will be empty, to accommodate a needle. For a completely filled column add suffix -901. Agilent configuration also includes 1.5 in. void on detector side.

Column Configuration Key: General - General Configuration

Specifications

Max. Temperature 290°C
Temperature Range -60°C to +290°C
Length (Metric) 1828.8 mm
Material Stainless Steel
Packing Material 3% Rtx™-5
Mesh Size 100/120 Silcoport W
Diameter (Metric) Inner 2 mm
Diameter (Metric) Outer 3.17 mm
USP Type G27, G36
Type Varian 3700, Vista Series, FID
Stationary Phase Bonded Phase 3% Rtx™-5
For Use With (Equipment) On-column Injections
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.