Learn More
Restek™ Capillary Column Caps
SDP
Catalog No. 06718381 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
06-718-381 10/Pk.
1 options
Catalog No. 06-718-381 Supplier Restek™ Supplier No. 21044
Attach to the column in seconds to form an airtight seal.
  • Increase column lifetime by preventing moisture and air from entering the column during storage
  • Made from glass
  • Color-coded for identifying detector and injector ends
  • Not recommended for reuse
Specifications
Type Column Caps
Quantity 10/Pk.