Learn More
Restek™ Capillary Column Rinsing Reservoir
SDP
Catalog No. 06710848 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Type:
Rinsing Reservoir
Rinsing Reservoir Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
06-710-848 Rinsing Reservoir
06-710-847 Rinsing Reservoir Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 06-710-848 Supplier Restek™ Supplier No. 20613
Restore the performance of bonded-phase capillary columns by dissolving and removing soluble, nonvolatile residue, using this reservoir kit.
  • 50mL rinsing reservoir equipped with 0.25 in. inlet and outlet connections
  • Built-in fritted disk prevents particulate matter from contaminating the column
  • Includes reservoir, pressure regulator, fittings, ferrules, and tubing
Order Info
Reservoir also available separately
Specifications
Type Rinsing Reservoir
Includes Rinsing Reservoir only
For Use With Capillary Column