Learn More
Restek™ Capillary Dedicated FID Jet for Agilent™ 6890/6850 GC
SDP
Catalog No. p-3181041 Shop All Restek Corporation Products
Change view
Click to view available options
Type:
High-Performance Siltek Treated
Standard
Packaging Quantity:
3 Pack
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Packaging Quantity
06-711-691 Standard Each
06-711-690 High-Performance Siltek Treated Each
06-711-723 Standard 3 Pack
06-711-724 High-Performance Siltek Treated 3 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 06-711-691 Supplier Restek™ Supplier No. 21621
Only null left
Add to Cart
Add to Cart
Fluted tip guides the capillary column into the jet
  • Threads specially coated for easy installation and removal
  • Standard version undergoes special processing ensures the highest degree of cleanliness
  • High-performance Siltek™ version is extremely inert for use with active compounds
Specifications
Type Standard
Packaging Quantity Each
For Use With Agilent 6890/6850