Learn More
Restek™ Capillary FID Jets for Agilent™ 5890/6890/6850 GC
SDP
Catalog No. p-3181029
Click to view available options
Type:
High-Performance Siltek Treated
Standard
Packaging Quantity:
3 Pack
Each
Fluted tip to guide the capillary column into the jet
  • Threads specially coated for easy installation and removal
  • Standard version undergoes special processing to ensure the highest degree of cleanliness
  • High-performance version is Siltek™ treated to render extremely inert for use with active compounds
Specifications
Type Standard
Packaging Quantity Each
For Use With Agilent 5890/6890/6850
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.