Learn More
Restek™ Capillary Installation Gauge for Agilent MS
SDP
Catalog No. 06711785 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Packaging Quantity:
1 Each
Catalog No. Packaging Quantity
06-711-785 1 Each
1 options
Catalog No. 06-711-785 Supplier Restek™ Supplier No. 21894
For use with Agilent 5973 and 5975 MS.
  • Made from high-quality stainless steel
  • Seats ferrules onto column for consistent installations
Specifications
Type Capillary Installation Gauge
For Use With (Application) Seats Ferrules Onto Column for Consistent Installations
Packaging Quantity 1 Each
For Use With Agilent 5973 and 5975 MS
Material Stainless Steel